5-Chlor-2-thiophencarbonylchlorid 97%
Lieferant: Apollo Scientific
|
Gefahr
|
OR200094-25G
APOSOR200094-25GEA
69.6
EUR
APOSOR200094-25G
5-Chlor-2-thiophencarbonylchlorid 97%
5-Chlorothiophene-2-carbonyl chloride
Formel:
C5H2Cl2OS Molecular Weight: 181,04 g/mol Schmelzpunkt: 4 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00043886 CAS-Nummer: 42518-98-9 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...