1-Bromo-4-(4-nitrophenoxy)benzene

Lieferant: Apollo Scientific

OR200100-1G OR200100-250MG OR200100-500MG OR200100-2.5G OR200100-5G
APOSOR200100-1GEA 314 EUR
APOSOR200100-1G APOSOR200100-250MG APOSOR200100-500MG APOSOR200100-2.5G APOSOR200100-5G
1-Bromo-4-(4-nitrophenoxy)benzene
1-Bromo-4-(4-nitrophenoxy)benzene
Formel: C₁₂H₈BrNO₃
Molecular Weight: 294.11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00222614
CAS-Nummer: 21969-04-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR