4-(4-Bromophenoxy)benzonitrile

Lieferant: Apollo Scientific

OR200101-1G OR200101-250MG OR200101-500MG OR200101-2.5G
APOSOR200101-1GEA 0 EUR
APOSOR200101-1G APOSOR200101-250MG APOSOR200101-500MG APOSOR200101-2.5G
4-(4-Bromophenoxy)benzonitrile
4-(4-Bromophenoxy)benzonitrile
Formel: C₁₃H₈BrNO
Lagertemperatur: Raumtemperatur
MDL: MFCD08446949
CAS-Nummer: 330792-93-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR