2-(4-Morpholino)-5-nitropyridin 98%

Lieferant: Apollo Scientific

Synonyme: 4-(5-Nitropyridin-2-yl)morpholine

OR200103-25G
APOSOR200103-25GEA 104 EUR
APOSOR200103-25G
2-(4-Morpholino)-5-nitropyridin 98%
2-(4-Morpholino)-5-nitropyridin
Formel: C₉H₁₁N₃O₃
Molecular Weight: 209.2019 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00023308
CAS-Nummer: 26820-62-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR