6-Chloro-4-methylpyridine-2-carboxylic acid 98%

Lieferant: Apollo Scientific

OR200109-5G
APOSOR200109-5GEA 59.6 EUR
APOSOR200109-5G
6-Chloro-4-methylpyridine-2-carboxylic acid 98%
6-Chloro-4-methylpyridine-2-carboxylic acid
Formel: C₇H₆ClNO₂
Molecular Weight: 171.58 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09834827
CAS-Nummer: 324028-95-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR