3-(Bromomethyl)-2-nitropyridine

Lieferant: Apollo Scientific

OR200119-250MG
APOSOR200119-250MGEA 473 EUR
APOSOR200119-250MG
3-(Bromomethyl)-2-nitropyridine
3-(Bromomethyl)-2-nitropyridine
Formel: C₆H₅BrN₂O₂
Molecular Weight: 217.02 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD14584511
CAS-Nummer: 15936-06-8
UN: 3265
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR