N-(2-Methoxyethyl)-N-Propylamin
Lieferant: Apollo Scientific
OR200127-1G
APOSOR200127-1GEA
0
EUR
APOSOR200127-1G
N-(2-Methoxyethyl)-N-Propylamin
N-(2-Methoxyethyl)-N-Propylamin
Formel:
C₆H₁₅NO Molecular Weight: 117,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00144830 CAS-Nummer: 43175-57-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...