3-(2-Methylphenyl)propionoyl chloride

Lieferant: Apollo Scientific
Gefahr

OR200130-1G OR200130-250MG OR200130-500MG
APOSOR200130-1GEA 0 EUR
APOSOR200130-1G APOSOR200130-250MG APOSOR200130-500MG
3-(2-Methylphenyl)propionoyl chloride
3-(2-Methylphenyl)propionoyl chloride
Formel: C₁₀H₁₁ClO
Lagertemperatur: Raumtemperatur
MDL: MFCD16739488
CAS-Nummer: 85829-29-4
UN: 3265
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR