(4-Hydroxy-1-piperidinyl)(3-piperidinyl)methanone hydrochloride

Lieferant: Apollo Scientific

Synonyme: (4-Hydroxypiperidin-1-yl)piperidin-3-ylmethanone

OR200135-1G
APOSOR200135-1GEA 0 EUR
APOSOR200135-1G
(4-Hydroxy-1-piperidinyl)(3-piperidinyl)methanone hydrochloride
(4-Hydroxy-1-piperidinyl)(3-piperidinyl)methanone hydrochloride
Formel: C₁₁H₂₁ClN₂O₂
Molecular Weight: 248,76 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06796403
CAS-Nummer: 496057-57-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR