5-Bromo-6-chloronicotinoyl chloride

Lieferant: Apollo Scientific

OR200143-1G OR200143-100MG OR200143-250MG OR200143-500MG OR200143-2.5G
APOSOR200143-1GEA 589 EUR
APOSOR200143-1G APOSOR200143-100MG APOSOR200143-250MG APOSOR200143-500MG APOSOR200143-2.5G
5-Bromo-6-chloronicotinoyl chloride
5-Bromo-6-chloronicotinoyl chloride
Formel: C₆H₂BrCl₂NO
Molecular Weight: 254.896 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16037365
CAS-Nummer: 78686-84-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR