1-(4-Methyl-4,5-dihydro-1H-imidazol-2-yl)-1H-pyrazole hydroiodide

Lieferant: Apollo Scientific

OR200146-1G
APOSOR200146-1GEA 451 EUR
APOSOR200146-1G
1-(4-Methyl-4,5-dihydro-1H-imidazol-2-yl)-1H-pyrazole hydroiodide
1-(4-Methyl-4,5-dihydro-1H-imidazol-2-yl)-1H-pyrazole hydroiodide
Formel: C₇H₁₀N₄·HI
Molecular Weight: 278,094 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28154661
CAS-Nummer: 1822851-86-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR