2-Bromopyridine-6-carbonyl chloride

Lieferant: Apollo Scientific

OR200166-1G OR200166-50MG OR200166-250MG OR200166-500MG
APOSOR200166-1GEA 0 EUR
APOSOR200166-1G APOSOR200166-50MG APOSOR200166-250MG APOSOR200166-500MG
2-Bromopyridine-6-carbonyl chloride
2-Bromopyridine-6-carbonyl chloride
Formel: C₆H₃BrClNO
Lagertemperatur: Raumtemperatur
MDL: MFCD18909527
CAS-Nummer: 139487-69-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR