3-(Acetylamino)benzoyl chloride
Lieferant: Apollo Scientific
Synonyme:
2,6-Dimethylnicotinoyl chloride
OR200167-250MG
APOSOR200167-250MGEA
249
EUR
APOSOR200167-250MG
3-(Acetylamino)benzoyl chloride
3-(Acetylamino)benzoyl chloride
Formel:
C₉H₈ClNO₂ Molecular Weight: 197,62 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11939183 CAS-Nummer: 90531-74-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...