5-(Aminomethyl)-1,3-thiazole hydrochloride 95%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Thiazol-5-ylmethanamine hydrochloride, (1,3-Thiazol-5-yl)methylamine hydrochloride
OR20040-1G
OR20040-5G
OR20040-10G
APOSOR20040-1GEA
106
EUR
APOSOR20040-1G
APOSOR20040-5G
APOSOR20040-10G
5-(Aminomethyl)-1,3-thiazole hydrochloride 95%
5-(Aminomethyl)-1,3-thiazole hydrochloride
Formel:
C₄H₆N₂S·ClH Schmelzpunkt: 173…181 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD06738808 CAS-Nummer: 131052-46-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...