5-(Aminomethyl)-1,3-thiazole hydrochloride 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: Thiazol-5-ylmethanamine hydrochloride, (1,3-Thiazol-5-yl)methylamine hydrochloride

OR20040-1G OR20040-5G OR20040-10G
APOSOR20040-1GEA 106 EUR
APOSOR20040-1G APOSOR20040-5G APOSOR20040-10G
5-(Aminomethyl)-1,3-thiazole hydrochloride 95%
5-(Aminomethyl)-1,3-thiazole hydrochloride
Formel: C₄H₆N₂S·ClH
Schmelzpunkt: 173…181 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06738808
CAS-Nummer: 131052-46-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR