4-Cyano-1H-indole-2-boronic acid, pinacol ester 95%
Lieferant: Apollo Scientific
OR20047-1G
OR20047-250MG
APOSOR20047-1GEA
307
EUR
APOSOR20047-1G
APOSOR20047-250MG
4-Cyano-1H-indole-2-boronic acid, pinacol ester 95%
4-Cyano-1H-indole-2-boronic acid, pinacol ester
Formel:
C₁₅H₁₇BN₂O₂ Lagertemperatur: Gefrierschrank |
MDL:
MFCD11858395 CAS-Nummer: 1256359-19-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...