5-Methylfuran-2-boronsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Methyl-2-furanboronic acid, 5-Methyl-2-furylboronic acid, (5-Methylfuran-2-yl)boronic acid

OR2011-1G OR2011-250MG OR2011-5G
APOSOR2011-1GEA 55.1 EUR
APOSOR2011-1G APOSOR2011-250MG APOSOR2011-5G
5-Methylfuran-2-boronsäure 97%
5-Methylfuran-2-boronsäure
Formel: C₅H₇BO₃
Molecular Weight: 125.9183 g/mol
Schmelzpunkt: 82 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01114644
CAS-Nummer: 62306-79-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR