2-Pyridincarbaldehydoxim

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Pyridinecarbaldehyde oxime, 2-Pyridinealdoxime

OR2013-5G
APOSOR2013-5GEA 50.9 EUR
APOSOR2013-5G
2-Pyridincarbaldehydoxim
2-Pyridincarbaldehydoxim
Formel: C₆H₆N₂O
Molecular Weight: 122,13 g/mol
Schmelzpunkt: 110…112 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006354
CAS-Nummer: 873-69-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR