(1H-Imidazol-4-yl)methanol Hydrochlorid 98+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Imidazolemethanol hydrochloride, 4(5)-Imidazolemethanol hydrochloride, 4-(Hydroxymethyl)imidazole hydrochloride

OR2014-1G OR2014-25G OR2014-5G
APOSOR2014-1GEA 21.9 EUR
APOSOR2014-1G APOSOR2014-25G APOSOR2014-5G
(1H-Imidazol-4-yl)methanol Hydrochlorid 98+%
(1H-Imidazol-4-yl)methanol Hydrochlorid
Formel: C₄H₇ClN₂O
Schmelzpunkt: 108…112 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00012697
CAS-Nummer: 32673-41-9
EINECS: 251-150-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR