1-(2,3-Dichlorphenyl)biguanid Hydrochlorid
Lieferant: Apollo Scientific
|
Achtung
|
OR2019-100G
OR2019-25G
OR2019-5G
APOSOR2019-100GEA
1080
EUR
APOSOR2019-100G
APOSOR2019-25G
APOSOR2019-5G
1-(2,3-Dichlorphenyl)biguanid Hydrochlorid
1-(2,3-Dichlorphenyl)biguanid Hydrochlorid
Formel:
C₈H₁₀Cl₃N₅ Schmelzpunkt: 228…232 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00053017 CAS-Nummer: 175205-08-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...