(±)-1-Pentin-3-ol 95%

Lieferant: Apollo Scientific

OR2042-5G
APOSOR2042-5GEA 0 EUR
APOSOR2042-5G
(±)-1-Pentin-3-ol 95%
(±)-1-Pentin-3-ol
Formel: C₅H₈O
Lagertemperatur: Raumtemperatur
MDL: MFCD00004572
CAS-Nummer: 4187-86-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR