1-(2-Iodobenzyl)-1H-1,2,4-triazole 97+%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1-Iodo-2-[(1H-1,2,4-triazol-1-yl)methyl]benzene, 1-[(2-Iodophenyl)methyl]-1H-1,2,4-triazole
OR2053-1G
APOSOR2053-1GEA
109
EUR
APOSOR2053-1G
1-(2-Iodobenzyl)-1H-1,2,4-triazole 97+%
1-(2-Iodobenzyl)-1H-1,2,4-triazole
Formel:
C₉H₈IN₃ Molecular Weight: 285,084 g/mol Schmelzpunkt: 67…69,5 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08741431 CAS-Nummer: 876316-31-3 UN: 1325 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...