5-(Hydroxymethyl)-1H-benzimidazole 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(1H-Benzimidazol-5-yl)methanol
OR2056-1G
OR2056-250MG
APOSOR2056-1GEA
255
EUR
APOSOR2056-1G
APOSOR2056-250MG
5-(Hydroxymethyl)-1H-benzimidazole 97%
5-(Hydroxymethyl)-1H-benzimidazole
Formel:
C₈H₈N₂O Schmelzpunkt: 129…131 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08690267 CAS-Nummer: 106429-29-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...