1-[4-(Aminomethyl)benzyl]-1H-pyrazole hydrochloride, tech

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(1H-Pyrazol-1-ylmethyl)benzylamine hydrochloride

OR2066-1G OR2066-250MG
APOSOR2066-1GEA 261 EUR
APOSOR2066-1G APOSOR2066-250MG
1-[4-(Aminomethyl)benzyl]-1H-pyrazole hydrochloride, tech
1-[4-(Aminomethyl)benzyl]-1H-pyrazole hydrochloride
Formel: C₁₁H₁₃N₃·ClH
Schmelzpunkt: 198…199,5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07690519
CAS-Nummer: 904696-62-4
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR