6-Phenylnicotinic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Phenylpyridine-3-carboxylic acid, 6-Phenylnicotinic acid

OR2073-250MG
APOSOR2073-250MGEA 21.9 EUR
APOSOR2073-250MG
6-Phenylnicotinic acid 97%
6-Phenylnicotinic acid
Formel: C₁₂H₉NO₂
Molecular Weight: 199,21 g/mol
Schmelzpunkt: 232…233 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03086128
CAS-Nummer: 29051-44-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR