2-(3-Methyl-1,2,4-oxadiazol-5-yl)benzoic acid

Lieferant: Apollo Scientific
Achtung

OR2076-1G OR2076-250MG
APOSOR2076-1GEA 275 EUR
APOSOR2076-1G APOSOR2076-250MG
2-(3-Methyl-1,2,4-oxadiazol-5-yl)benzoic acid
2-(3-Methyl-1,2,4-oxadiazol-5-yl)benzoic acid
Formel: C₁₀H₈N₂O₃
Schmelzpunkt: 151…153 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08741426
CAS-Nummer: 475105-77-2
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR