2-Chlor-5-nitrozimtsäure 97%

Lieferant: Apollo Scientific

Synonyme: 2-Chlor-5-nitrocinnamsäure, 3-(2-Chlor-5-nitrophenyl)propensäure

OR2084-25G
APOSOR2084-25GEA 132 EUR
APOSOR2084-25G
2-Chlor-5-nitrozimtsäure 97%
2-Chlor-5-nitrozimtsäure
Formel: C₉H₆ClNO₄
Molecular Weight: 227.6 g/mol
Schmelzpunkt: 219…221 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00063312
CAS-Nummer: 36015-19-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR