2,5-Dibrom-4-picolin
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2,5-Dibrom-4-methylpyridin
OR2087-100G
OR2087-25G
OR2087-5G
APOSOR2087-100GEA
139
EUR
APOSOR2087-100G
APOSOR2087-25G
APOSOR2087-5G
2,5-Dibrom-4-picolin
2,5-Dibrom-4-picolin
Formel:
C₆H₅Br₂N Schmelzpunkt: 36…42 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00234955 CAS-Nummer: 3430-26-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...