2-(3-Nitrophenoxy)pyridine 97+%

Lieferant: Apollo Scientific

OR2092-1G OR2092-5G
APOSOR2092-1GEA 112 EUR
APOSOR2092-1G APOSOR2092-5G
2-(3-Nitrophenoxy)pyridine 97+%
2-(3-Nitrophenoxy)pyridine
Formel: C₁₁H₈N₂O₃
Molecular Weight: 216.2 g/mol
Schmelzpunkt: 93…95 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08690283
CAS-Nummer: 28355-48-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR