2-Methylbenzothiazole-6-sulphonyl chloride 95+%

Lieferant: Apollo Scientific
Gefahr

OR2093-250MG
APOSOR2093-250MGEA 132 EUR
APOSOR2093-250MG
2-Methylbenzothiazole-6-sulphonyl chloride 95+%
2-Methylbenzothiazole-6-sulphonyl chloride
Formel: C₈H₆ClNO₂S₂
Molecular Weight: 247.72 g/mol
Schmelzpunkt: 98…101 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03990571
CAS-Nummer: 21431-13-0
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR