4-Chloro-5-(2-thienyl)thieno[2,3-d]pyrimidine

Lieferant: Apollo Scientific

OR21001-1G
APOSOR21001-1GEA 145 EUR
APOSOR21001-1G
4-Chloro-5-(2-thienyl)thieno[2,3-d]pyrimidine
4-Chloro-5-(2-thienyl)thieno[2,3-d]pyrimidine
Formel: C₁₀H₅ClN₂S₂
Molecular Weight: 252,734 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00174019
CAS-Nummer: 189681-04-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR