2-Bromo-N-(2,6-dimethylphenyl)propanamide
Lieferant: Apollo Scientific
|
Achtung
|
OR21004-100MG
APOSOR21004-100MGEA
21.9
EUR
APOSOR21004-100MG
2-Bromo-N-(2,6-dimethylphenyl)propanamide
2-Bromo-N-(2,6-dimethylphenyl)propanamide
Formel:
C₁₁H₁₄BrNO Schmelzpunkt: 163…165 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00224833 CAS-Nummer: 41708-73-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...