6-Brom-4-chinazolon

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Bromo-4(3H)-quinazolinone

OR2103-1G
APOSOR2103-1GEA 21.9 EUR
APOSOR2103-1G
6-Brom-4-chinazolon
6-Brom-4-chinazolon
Formel: C₈H₅BrN₂O
Molecular Weight: 225,04 g/mol
Schmelzpunkt: 267…271 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01219541
CAS-Nummer: 32084-59-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR