3-Imidazo[2,1-b]thiazol-6-ylphenylamine

Lieferant: Apollo Scientific
Achtung

OR2104-1G
APOSOR2104-1GEA 145 EUR
APOSOR2104-1G
3-Imidazo[2,1-b]thiazol-6-ylphenylamine
3-Imidazo[2,1-b]thiazol-6-ylphenylamine
Formel: C₁₁H₉N₃S
Molecular Weight: 215,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03617826
CAS-Nummer: 861206-26-0
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR