N-(4-Amino-2-cyanophenyl)acetamide

Lieferant: Apollo Scientific
Gefahr

OR21114-1G
APOSOR21114-1GEA 386 EUR
APOSOR21114-1G
N-(4-Amino-2-cyanophenyl)acetamide
N-(4-Amino-2-cyanophenyl)acetamide
Formel: C₉H₉N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD00115481
CAS-Nummer: 73894-39-0
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR