4-Hydroxychinaldin 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Methyl-4-chinolinol
OR21133-25G
APOSOR21133-25GEA
174
EUR
APOSOR21133-25G
4-Hydroxychinaldin 98%
4-Hydroxychinaldin
Formel:
C₁₀H₉NO Molecular Weight: 159,19 g/mol Schmelzpunkt: 234…236 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006758 CAS-Nummer: 607-67-0 EINECS: 210-140-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...