4-(4-Pyridylmethyl)aniline

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(4-pyridinylmethyl)benzenamine, 4-(Pyridin-4-ylmethyl)aniline

OR21140-1G
APOSOR21140-1GEA 98.6 EUR
APOSOR21140-1G
4-(4-Pyridylmethyl)aniline
4-(4-Pyridylmethyl)aniline
Formel: C₁₂H₁₂N₂
Molecular Weight: 184,237 g/mol
Schmelzpunkt: 156…160 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00053060
CAS-Nummer: 27692-74-6
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR