2-Amino-5-methylbenzohydrazide

Lieferant: Apollo Scientific
Achtung

OR21156-250MG
APOSOR21156-250MGEA 29 EUR
APOSOR21156-250MG
2-Amino-5-methylbenzohydrazide
2-Amino-5-methylbenzohydrazide
Formel: C₈H₁₁N₃O
Molecular Weight: 165.2 g/mol
Schmelzpunkt: 133…134 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00111324
CAS-Nummer: 28461-49-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR