8-Nitroimidazo[1,2-a]pyridin

Lieferant: Apollo Scientific

OR2116-1G
APOSOR2116-1GEA 24.4 EUR
APOSOR2116-1G
8-Nitroimidazo[1,2-a]pyridin
8-Nitroimidazo[1,2-a]pyridin
Formel: C₇H₅N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08056674
CAS-Nummer: 52310-46-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR