2-Methyl-N-[2-(pyridin-2-yl)ethyl]aniline

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-{2-[(2-Methylphenyl)amino]ethyl}pyridine, 2-{[2-(Pyridin-2-yl)ethyl]amino}toluene

OR21161-100MG
APOSOR21161-100MGEA 50.9 EUR
APOSOR21161-100MG
2-Methyl-N-[2-(pyridin-2-yl)ethyl]aniline
2-Methyl-N-[2-(pyridin-2-yl)ethyl]aniline
Formel: C₁₄H₁₆N₂
Molecular Weight: 212,29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00023534
CAS-Nummer: 92733-82-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR