1-(4-Nitrophenyl)pyrrolidin

Lieferant: Apollo Scientific
Achtung

OR2117-1G
APOSOR2117-1GEA 21.9 EUR
APOSOR2117-1G
1-(4-Nitrophenyl)pyrrolidin
1-(4-Nitrophenyl)pyrrolidin
Formel: C₁₀H₁₂N₂O₂
Molecular Weight: 192,214 g/mol
Schmelzpunkt: 163…166 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00020819
CAS-Nummer: 10220-22-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR