3-(1-Pyrrolyl)benzonitril
Lieferant: Apollo Scientific
OR21182-1G
APOSOR21182-1GEA
0
EUR
APOSOR21182-1G
3-(1-Pyrrolyl)benzonitril
3-(1-Pyrrolyl)benzonitril
Formel:
C₁₁H₈N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052877 CAS-Nummer: 175134-98-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...