2-(4-Nitrophenyl)-1,3,4-oxadiazole

Lieferant: Apollo Scientific

OR2119-1G
APOSOR2119-1GEA 0 EUR
APOSOR2119-1G
2-(4-Nitrophenyl)-1,3,4-oxadiazole
2-(4-Nitrophenyl)-1,3,4-oxadiazole
Formel: C₈H₅N₃O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00491617
CAS-Nummer: 4291-13-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR