N1-(8-quinolyl)-2-chloroacetamide
Lieferant: Apollo Scientific
OR21196-1G
APOSOR21196-1GEA
226
EUR
APOSOR21196-1G
N1-(8-quinolyl)-2-chloroacetamide
N1-(8-quinolyl)-2-chloroacetamide
Formel:
C₁₁H₉ClN₂O Molecular Weight: 220,655 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00218186 CAS-Nummer: 32889-11-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...