1-Benzenesulphonyl-4-chloro-2-nitrobenzene
Lieferant: Apollo Scientific
|
Gefahr
|
OR21208-250MG
APOSOR21208-250MGEA
29
EUR
APOSOR21208-250MG
1-Benzenesulphonyl-4-chloro-2-nitrobenzene
1-Benzenesulphonyl-4-chloro-2-nitrobenzene
Formel:
C₁₂H₈ClNO₄S Schmelzpunkt: 121 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052250 CAS-Nummer: 86030-08-2 UN: 1759 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...