ω-Brom-3,4-dichloracetophenon
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
α-Brom-3,4-dichloracetophenon, 2-Brom-1-(3,4-dichlorphenyl)ethan-1-on, 3,4-Dichlorphenacylbromid, 2-Brom-3',4'-dichloracetophenon
OR21212-5G
OR21212-1G
OR21212-2.5G
OR21212-10G
APOSOR21212-5GEA
65.4
EUR
APOSOR21212-5G
APOSOR21212-1G
APOSOR21212-2.5G
APOSOR21212-10G
ω-Brom-3,4-dichloracetophenon
ω-Brom-3,4-dichloracetophenon
Formel:
C₈H₅BrCl₂O Molecular Weight: 267.94 g/mol Schmelzpunkt: 55…60 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00051581 CAS-Nummer: 2632-10-2 UN: 2923 ADR: 8+6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...