2-Cyan-5-methoxybenzoesäure

Lieferant: Apollo Scientific
Achtung

OR21216-100MG
APOSOR21216-100MGEA 55.1 EUR
APOSOR21216-100MG
2-Cyan-5-methoxybenzoesäure
2-Cyano-5-methoxybenzoic acid
Formel: C₉H₇NO₃
Molecular Weight: 177,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00116264
CAS-Nummer: 179028-65-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR