6-Methoxy-o-kresol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Hydroxy-3-methylanisol, 1-Hydroxy-2-methoxy-6-methylbenzol, 2-Hydroxy-3-methoxytoluol, 2-Methoxy-6-methylphenol

OR21227-1G
APOSOR21227-1GEA 21.9 EUR
APOSOR21227-1G
6-Methoxy-o-kresol 98%
6-Methoxy-o-kresol
Formel: C₈H₁₀O₂
Molecular Weight: 138,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00043771
CAS-Nummer: 2896-67-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR