Ethyl-4-amino-3,5-dichlorbenzoat

Lieferant: Apollo Scientific
Achtung

OR21247-1G
APOSOR21247-1GEA 72.5 EUR
APOSOR21247-1G
Ethyl-4-amino-3,5-dichlorbenzoat
Ethyl-4-amino-3,5-dichlorbenzoat
Formel: C₉H₉Cl₂NO₂
Molecular Weight: 234,793 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00016636
CAS-Nummer: 74878-31-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR