5-Methyl-2-phenyl-2H-1,2,3-triazole-4-carbohydrazide

Lieferant: Apollo Scientific

OR21249-1G
APOSOR21249-1GEA 226 EUR
APOSOR21249-1G
5-Methyl-2-phenyl-2H-1,2,3-triazole-4-carbohydrazide
5-Methyl-2-phenyl-2H-1,2,3-triazole-4-carbohydrazide
Formel: C₁₀H₁₁N₅O
Molecular Weight: 217,227 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00052546
CAS-Nummer: 175135-03-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR